Läs mer
Tetrabromobisphenol A-13C12, TRC
Produktkod. 30425255
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425255 10 mg
30420204 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425255 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29110810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,3',5,5'-Tetrabromobisphenol A ring-13C12
CAS 1352876-39-1
Anteckningar om renhetsgrad HPLC
Molekylformel 13C12 C3 H12 Br4 O2
Kvantitet 10 mg
Synonym Phenol-1,2,3,4,5,6-13C6','4,4'-(1-methylethylidene)bis[2,6-dibromo-, 4,4'-(2,2-Propanediyl)bis(2,6-dibromo-1,2,3,4,5,6-13C6-phenol), 2,2',6,6'-Tetrabromobisphenol A (ring-13C12), 3,3',5,5'-Tetrabromobisphenol A (ring-13C12)
InChI formel InChI=1S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3/i3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1
LEDER CC([13C]1=[13CH][13C](Br)=[13C](O)[13C](Br)=[13CH]1)(C)[13C]2=[13CH][13C](Br)=[13C](O)[13C](Br)=[13CH]2
IUPAC-namn 2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)propan-2-yl](1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
Molekylvikt (g/mol) 555.78
Formel vikt 551.7973