Läs mer
Tetrabromobisphenol S Dimethyl Ether, TRC
Produktkod. 30412122
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30412122 2.5 g 2.5g
30426792 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412122 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2911152.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70156-79-5
Molekylformel C14H10Br4O4S
Molekylvikt (g/mol) 593.91
Synonym Tetrabromobisphenol S-Dimethyl Ether, 1,1'-Sulfonylbis[3,5-dibromo-4-methoxybenzene], (3,5-Dibromo-4-methoxyphenyl) sulfone, 1,3-Dibromo-5-(3,5-dibromo-4-methoxybenzenesulfonyl)-2-methoxybenzene, Tetrabromobisphenol S dimethyl ether
IUPAC-namn 1,3-dibromo-5-(3,5-dibromo-4-methoxyphenyl)sulfonyl-2-methoxybenzene
LEDER COc1c(Br)cc(cc1Br)S(=O)(=O)c2cc(Br)c(OC)c(Br)c2
Specifikationer
Kemiskt namn eller material Tetrabromobisphenol S Dimethyl Ether
Kvantitet 2.5 g
InChI formel InChI=1S/C14H10Br4O4S/c1-21-13-9(15)3-7(4-10(13)16)23(19,20)8-5-11(17)14(22-2)12(18)6-8/h3-6H,1-2H3
Formel vikt 589.7033