Learn More
Thermo Scientific Chemicals Tetrabromophenol Blue
Catalog Number p-7024877
Click to view available options
Quantity:
1 g
5 g
25 g
4430-25-5
C19H6Br8O5S
985.544
MFCD00005876
QPMIVFWZGPTDPN-UHFFFAOYSA-N
3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
78159
2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br
4430-25-5
C19H6Br8O5S
985.544
MFCD00005876
QPMIVFWZGPTDPN-UHFFFAOYSA-N
3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
78159
2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY
Melting Point 203°C to 205°C (decomposition)
Beilstein 378438
Solubility Information Soluble in water.
Formula Weight 985.55
Quantity 1 g
Chemical Name or Material Tetrabromophenol Blue