Läs mer
Tetrabromoterephthalic Acid, TRC
Produktkod. 30410239
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30410239 5 g
1 options
Produktkod. 30410239 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2911355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetrabromoterephthalic Acid
CAS 5411-70-1
Molekylformel C8H2Br4O4
Kvantitet 5 g
Synonym 2,3,5,6-Tetrabromo-1,4-benzenedicarboxylic Acid, NSC 12442, Tetrabromo-1,4-benzenedicarboxylic Acid, Tetrabromoterephthalic Acid
InChI formel InChI=1S/C8H2Br4O4/c9-3-1(7(13)14)4(10)6(12)2(5(3)11)8(15)16/h(H,13,14)(H,15,16)
LEDER CC(N(OC(C)=O)C(N)=O)C(S1)=CC2=C1C=CC=C2
Molekylvikt (g/mol) 481.72