Läs mer
Tetrabutylammonium Dibromochloride, TRC
Produktkod. 30456802
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30456802 500 mg
1 options
Produktkod. 30456802 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT310783500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetrabutylammonium Dibromochloride
CAS 64531-21-1
Molekylformel C16H36Br2ClN
Kvantitet 500 mg
Synonym 1-Butanaminium','N,N,N-tributyl-','bromochlorobromate(1-) (9CI), Bromate(1-)','bromochloro-','N,N,N-tributyl-1-butanaminium (9CI), N,N,N,N-Tetrabutylammonium dibromochloride, Tetrabutylammonium dibromochloride
InChI formel InChI=1S/C16H36N.Br2Cl/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2-3/h5-16H2,1-4H3;/q+1;-1
LEDER CCCC[N+](CCCC)(CCCC)CCCC.Br[Br-]Cl
Molekylvikt (g/mol) 437.72
Formel vikt 435.0903