Läs mer
Tetrabutylammonium Diphenylphosphinate, TRC
Produktkod. 30419258
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419258 25 g
30425815 10 g
30428147 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30419258 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29125525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 208337-00-2
Molekylformel C28H46NO2P
Molekylvikt (g/mol) 459.64
Synonym N,N,N-Tributyl-1-butanaminium Diphenylphosphinate, Diphenylphosphinic Acid Ion(1-) N,N,N-Tributyl-1-butanaminium
IUPAC-namn diphenylphosphinate;tetrabutylazanium
LEDER CCCC[N+](CCCC)(CCCC)CCCC.[O-]P(=O)(c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Tetrabutylammonium Diphenylphosphinate
Kvantitet 25 g
InChI formel InChI=1S/C16H36N.C12H11O2P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h5-16H2,1-4H3;1-10H,(H,13,14)/q+1;/p-1
Formel vikt 459.3266