missing translation for 'onlineSavingsMsg'
Läs mer

Tetracaine Hydrochloride, TRC

Product Code. 30420338
Change view
Click to view available options
Kvantitet:
50 mg
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30420338 50 mg
30418636 25 g
30428425 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420338 Supplier Toronto Research Chemicals Supplier No. TRCT29133550MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-47-0
Molekylformel C15 H24 N2 O2 . Cl H
Molekylvikt (g/mol) 300.82
Synonym Tetracaine Hydrochloride, Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Benzoic acid, p-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Ethanol, 2-dimethylamino-, p-butylaminobenzoate, HCl (4CI), 2-(Dimethylamino)ethyl p-(butylamino)benzoate hydrochloride, 2-Dimethylaminoethanol 4-n-butylaminobenzoate hydrochloride, Amethocaine hydrochloride, Amethocaine monohydrochloride, Anacel, Anethain, Anethaine, Butethanol, Curtacain, Decicain, Decicaine, Dessicain, Dianguang, Dicaine, Dikain, Intercaine, Leocaine, Menonasal, Niphanoid, Pantocain, Pantocaine, Pantocaine hydrochloride, Pantocaine monohydrochloride, Pontocaine hydrochloride, Tetocaine, Tetracaine chlorhydrate, Tetracaine hydrochloride, Tetracaine monohydrochloride, Tetracainium chloride, Tonexol, p-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester hydrochloride
IUPAC-namn 2-(dimethylamino)ethyl 4-(butylamino)benzoate;hydrochloride
LEDER Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C

Specifications

Kemiskt namn eller material Tetracaine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H
Formel vikt 300.1605
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.