Läs mer
Tetracaine Hydrochloride, TRC
Produktkod. 30428425
Change view
Kvantitet:
50 mg
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428425 10 g
30420338 50 mg
30418636 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428425 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29133510G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136-47-0
Molekylformel C15 H24 N2 O2 . Cl H
Molekylvikt (g/mol) 300.82
Synonym Tetracaine Hydrochloride, Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Benzoic acid, p-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Ethanol, 2-dimethylamino-, p-butylaminobenzoate, HCl (4CI), 2-(Dimethylamino)ethyl p-(butylamino)benzoate hydrochloride, 2-Dimethylaminoethanol 4-n-butylaminobenzoate hydrochloride, Amethocaine hydrochloride, Amethocaine monohydrochloride, Anacel, Anethain, Anethaine, Butethanol, Curtacain, Decicain, Decicaine, Dessicain, Dianguang, Dicaine, Dikain, Intercaine, Leocaine, Menonasal, Niphanoid, Pantocain, Pantocaine, Pantocaine hydrochloride, Pantocaine monohydrochloride, Pontocaine hydrochloride, Tetocaine, Tetracaine chlorhydrate, Tetracaine hydrochloride, Tetracaine monohydrochloride, Tetracainium chloride, Tonexol, p-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester hydrochloride
IUPAC-namn 2-(dimethylamino)ethyl 4-(butylamino)benzoate;hydrochloride
LEDER Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C
Specifikationer
Kemiskt namn eller material Tetracaine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H
Formel vikt 300.1605
Procent renhet >95%