Läs mer
Tetrachlorothiophene, TRC
Produktkod. 30423510
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30423510 250 mg
1 options
Produktkod. 30423510 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT292325250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,3,4,5-Tetrachlorothiophene
CAS 6012-97-1
Molekylformel C4 Cl4 S
Kvantitet 250 mg
Synonym Thiophene','2,3,4,5-tetrachloro- (ACI), 2,3,4,5-Tetrachlorothiophene (ACI), Thiophene','tetrachloro- (6CI','7CI','8CI','9CI), IF, NSC 44615, NSC 61442, Penphene, Perchlorothiophene, Tetrachlorothiophene
InChI formel InChI=1S/C4Cl4S/c5-1-2(6)4(8)9-3(1)7
LEDER Clc1sc(Cl)c(Cl)c1Cl
IUPAC-namn 2,3,4,5-tetrachlorothiophene
Molekylvikt (g/mol) 221.92
Formel vikt 219.8475