Läs mer
Tetracosyl Acetate, TRC
Produktkod. 30450988
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30450988 100 mg
1 options
Produktkod. 30450988 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT724485100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetracosyl Acetate
CAS 822-29-7
Molekylformel C26 H52 O2
Kvantitet 100 mg
Synonym Tetracosan-1-yl Acetate, Tetracosanol Acetate
InChI formel InChI=1S/C26H52O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-28-26(2)27/h3-25H2,1-2H3
LEDER CCCCCCCCCCCCCCCCCCCCCCCCOC(=O)C
IUPAC-namn tetracosyl acetate
Molekylvikt (g/mol) 396.69
Formel vikt 396.397