missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tetradecanedioinsyra, TRC
Specifications
Specifications
| Kemiskt namn eller material | 1,12-Dodecanedicarboxylic acid |
| CAS | 821-38-5 |
| Molekylformel | C14 H26 O4 |
| Kvantitet | 500 mg |
| Synonym | Tetradecanedioic acid (6CI','7CI','8CI','9CI','ACI), 1,12-Dicarboxydodecane, 1,12-Dodecanedicarboxylic acid, 1,14-Tetradecanedioic acid, DCA 14, Emerox 114, NSC 9504 |
| InChI formel | InChI=1S/C14H26O4/c15-13(16)11-9-7-5-3-1-2-4-6-8-10-12-14(17)18/h1-12H2,(H,15,16)(H,17,18) |
| LEDER | OC(=O)CCCCCCCCCCCCC(=O)O |
| IUPAC-namn | tetradekandisyra |
| Molekylvikt (g/mol) | 258.35 |
| Formel vikt | 258.1831 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?