Innehåller
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Tetradifon | 116-29-0 | 1000μg/mL |
Certifikat
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Uppfyllande
GHS/OSHA Complaint
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tetradifon |
| Förpackning | 2mL amber prescored glass ampule |
| CAS | 116-29-0, 67-56-1 |
| CAS Min % | 0.1, 99.9 |
| Anteckningar om koncentration eller sammansättning | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS Max % | 0.1, 99.9 |
| Molekylformel | C12H6Cl4O2S |
| För användning med (utrustning) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI-nyckel | MLGCXEBRWGEOQX-UHFFFAOYSA-N |
| LEDER | C1=CC(=CC=C1S(=O)(=O)C2=CC(=C(C=C2Cl)Cl)Cl)Cl |
| Visa mer |
Product intended for laboratory use only