Läs mer
Tetraethylammonium Cyanide, TRC
Produktkod. 30414437
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30414437 500 mg
1 options
Produktkod. 30414437 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT292715500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetraethylammonium Cyanide
CAS 13435-20-6
Molekylformel C8H20N.CN
Kvantitet 500 mg
Synonym N,N,N-Triethylethanaminium Cyanide, Tetraethylammonium Cyanide, N,N,N-triethylethanaminium Hydrocyanic Acid
InChI formel InChI=1S/C8H20N.CN/c1-5-9(6-2,7-3)8-4;1-2/h5-8H2,1-4H3;/q+1;-1
LEDER CC[N+](CC)(CC)CC.[C-]#N
IUPAC-namn tetraethylazanium;cyanide
Molekylvikt (g/mol) 156.27
Formel vikt 156.1626