Läs mer
Tetraetylammonium P-toluensulfonat, TRC
Specifications
Specifications
| Kemiskt namn eller material | Tetraethylammonium P-Toluenesulfonate |
| CAS | 733-44-8 |
| Molekylformel | C15H27NO3S |
| Kvantitet | 500 mg |
| Synonym | Ethanaminium','N,N,N-triethyl-','4-methylbenzenesulfonate (1:1) (ACI), Ammonium','tetraethyl-','p-toluenesulfonate (8CI), Ethanaminium','N,N,N-triethyl-','salt with 4-methylbenzenesulfonic acid (1:1) (9CI), Tetraethylammonium p-toluenesulfonate (6CI','7CI), Tetraethylammonium p-toluenesulphonate, Tetraethylammonium p-tolylsulfonate, Tetraethylammonium p-tosylate, Tetraethylammonium toluenesulfonate, Tetraethylammonium tosylate, N,N,N-Triethylethanaminium 4-methylbenzenesulfonate |
| InChI formel | InChI=1S/C8H20N.C7H8O3S/c1-5-9(6-2,7-3)8-4;1-6-2-4-7(5-3-6)11(8,9)10/h5-8H2,1-4H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1 |
| LEDER | CC[N+](CC)(CC)CC.Cc1ccc(cc1)S(=O)(=O)[O-] |
| IUPAC-namn | 4-metylbensensulfonat; tetraetylazanium |
| Molekylvikt (g/mol) | 301.45 |
| Formel vikt | 301.1712 |