Läs mer
Tetraethylcystamine, TRC
Produktkod. 30424519
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30424519 50 mg
1 options
Produktkod. 30424519 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29143550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetraethylcystamine
CAS 589-32-2
Molekylformel C12 H28 N2 S2
Kvantitet 50 mg
Synonym Ethanamine','2,2'-dithiobis[N,N-diethyl-, Triethylamine','2,2'''-dithiobis- (6CI,7CI,8CI), 2,2'-Dithiobis[N,N-diethylethanamine], 2,2'''-Dithiobistriethylamine, 6,7-Dithia-3,10-diazadodecane','3,10-diethyl-, N,N,N',N'-Tetraethylcystamine, NSC 529154, Tetraethylcystamine, 2,2'-(Disulfane-1,2-diyl)bis(N,N-diethylethanamine)
InChI formel InChI=1S/C12H28N2S2/c1-5-13(6-2)9-11-15-16-12-10-14(7-3)8-4/h5-12H2,1-4H3
LEDER CCN(CC)CCSSCCN(CC)CC
IUPAC-namn 2-[2-(diethylamino)ethyldisulfanyl]-N,N-diethylethanamine
Molekylvikt (g/mol) 264.49
Formel vikt 264.1694