missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl methylenediphosphonate, 97%, Thermo Scientific Chemicals

Catalog Number p-3830548
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1660-94-2
C9H22O6P2
288.22
MFCD00039887
STJWVOQLJPNAQL-UHFFFAOYSA-N
tetraethyl methylenediphosphonate, tetraethyl methanediphosphonate, tetraethyl diphosphonomethane, methylenebis diethylphosphonate, tetraethyl methylenebisphosphonate, phosphonic acid, methylenebis-, tetraethyl ester, methylenebis diethoxyphosphine oxide, methylenebisphosphonic acid, tetraethyl ester, phosphonic acid, methylenedi-, tetraethyl ester, methylenedi phosphonic acid tetraethyl ester
15455
1-[diethoxyphosphorylmethyl(ethoxy)phosphoryl]oxyethane
CCOP(=O)(CP(=O)(OCC)OCC)OCC
This item is not returnable. View return policy
1660-94-2
C9H22O6P2
288.22
MFCD00039887
STJWVOQLJPNAQL-UHFFFAOYSA-N
tetraethyl methylenediphosphonate, tetraethyl methanediphosphonate, tetraethyl diphosphonomethane, methylenebis diethylphosphonate, tetraethyl methylenebisphosphonate, phosphonic acid, methylenebis-, tetraethyl ester, methylenebis diethoxyphosphine oxide, methylenebisphosphonic acid, tetraethyl ester, phosphonic acid, methylenedi-, tetraethyl ester, methylenedi phosphonic acid tetraethyl ester
15455
1-[diethoxyphosphorylmethyl(ethoxy)phosphoryl]oxyethane
CCOP(=O)(CP(=O)(OCC)OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Brown-Yellow to Yellow
Density 1.1600g/mL
Boiling Point 171.0°C to 174.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2[P(O)(OC2H5)2]2
Refractive Index 1.4405 to 1.4445
Quantity 1 mL
Specific Gravity 1.16
Formula Weight 288.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tetraethyl methylenediphosphonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'