Läs mer
Tetrahydro 11-Deoxycorticosterone, TRC
Produktkod. 30135567
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30135567 50 mg
17911915 10 mg
17914285 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135567 Supplier Toronto Research Chemicals Supplier No. TRCT29327050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 567-03-3
Molekylformel C21 H34 O3
Molekylvikt (g/mol) 334.49
Synonym Pregnan-20-one, 3,21-dihydroxy-, (3α,5β)-, 5β-Pregnan-20-one, 3α,21-dihydroxy- (8CI), Corticosterone, 11-deoxytetrahydro- (6CI), Corticosterone, deoxytetrahydro- (7CI), (3α,5β)-3,21-Dihydroxypregnan-20-one, 21-Hydroxypregnanolone, 3α,21-Dihydroxy-5β-pregnan-20-one, 3α,5β-Tetrahydrodeoxycorticosterone, 5β-Pregnan-3α,21-diol-20-one, 5β-Pregnane-3α,21-diol-20-one, Deoxytetrahydrocorticosterone, Tetrahydro DOC, Tetrahydro-11-deoxycorticosterone, Tetrahydrodeoxycorticosterone
IUPAC-namn 2-hydroxy-1-[(3R,5R,8R,9S,10S,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
LEDER C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@@]4(C)CC[C@H]23
Specifications
Kemiskt namn eller material Tetrahydro 11-Deoxycorticosterone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C21H34O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h13-18,22-23H,3-12H2,1-2H3/t13-,14-,15+,16+,17+,18-,20+,21+/m1/s1
Formel vikt 334.2508
Procent renhet >95%