missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-pyran-4-one, 97%, Thermo Scientific™

Catalog Number p-4510430
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
29943-42-8
C5H8O2
100.117
JMJRYTGVHCAYCT-UHFFFAOYSA-N
tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
121599
oxan-4-one
C1COCCC1=O
This item is not returnable. View return policy
29943-42-8
C5H8O2
100.117
JMJRYTGVHCAYCT-UHFFFAOYSA-N
tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
121599
oxan-4-one
C1COCCC1=O
Density 1.08g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material Tetrahydro-4H-pyran-4-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'