Learn More
Tetrahydro-4H-pyran-4-one, 99% Product Code.: 10655262
Tetrahydro-4H-pyran-4-one, 99%
Product Code. p-4509654 Shop All Thermo Scientific Chemicals Products
5 g, Glass bottle, 5g
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. 10655262
Brand: Thermo Scientific Acros 189440050
Request Bulk or Custom Format
CAS: 29943-42-8 | C5H8O2 | 100.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29943-42-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
InChI Key JMJRYTGVHCAYCT-UHFFFAOYSA-N
Synonym tetrahydro-4h-pyran-4-one, 4-oxotetrahydropyran, tetrahydropyran-4-one, dihydro-2h-pyran-4 3h-one, 4h-pyran-4-one, tetrahydro, 4-thp, tetrahydro-2h-pyran-4-one, tetrahydro-pyran-4-one, 4-oxotetrahydopyran, tetrahydro-.gamma.-pyrone
PubChem CID 121599
IUPAC Name oxan-4-one
SMILES C1COCCC1=O
Specifications
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 166.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Quantity 5 g
Beilstein 17,I,131
Specific Gravity 1.08
Solubility Information Solubility in water: miscible
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrahydro-4H-pyran-4-one
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0441271 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code189440050,189440250
Lot NumberA0441271 Certificate TypeCertificate of Analysis Date19/04/2024 Product Code189440050,189440250
Lot NumberA0399584 Certificate TypeCertificate of Analysis Date15/12/2022 Product Code189440050,189440250
Lot NumberA0395141 Certificate TypeCertificate of Analysis Date15/12/2022 Product Code189440050,189440250
Lot NumberA0433710 Certificate TypeCertificate of Analysis Date05/09/2022 Product Code189440050,189440250
compliance-icons
Product Identifier
  • tetrahydro-4H-pyran-4-one
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Content Correction
Product Title
Tetrahydro-4H-pyran-4-one, 99% >