Läs mer
Tetrahydroabietic Acid (Stereoisomer), TRC
Produktkod. 30423832
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423832 25 mg
30426574 10 mg
30416773 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29447025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 358976-89-3
Molekylformel C20H34O2
Molekylvikt (g/mol) 306.48
Synonym Tetrahydro-abietic Acid, (1R,4aR,4bS,10aR)-Tetradecahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic Acid, (1R,4aR,4bS,10aR)-7-Isopropyl-1,4a-dimethyltetradecahydrophenanthrene-1-carboxylic Acid
IUPAC-namn (1R,4aR,4bS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,7,8,8a,9,10,10a-dodecahydrophenanthrene-1-carboxylic acid
LEDER C[C@@]12[C@](CCC3[C@]2([H])CCC(C(C)C)C3)([H])[C@@](C(O)=O)(C)CCC1
Specifikationer
Kemiskt namn eller material Tetrahydroabietic Acid (Stereoisomer)
Kvantitet 25 mg
InChI formel InChI=1S/C20H34O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h13-17H,5-12H2,1-4H3,(H,21,22)/t14?,15?,16-,17+,19+,20+/m0/s1
Formel vikt 306.2559