Läs mer
Tetrahydroabietyl Alcohol, TRC
Produktkod. 30179159
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179159 5 mg
30179531 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2930455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13393-93-6
Molekylformel C20 H36 O
Molekylvikt (g/mol) 292.5
Synonym 1-Phenanthrenemethanol, tetradecahydro-1,4a-dimethyl-7-(1-methylethyl)-, Abietyl alcohol, tetrahydro- (6CI), (1R,4aR,4bS,10aR)-Tetradecahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenemethanol, Tetrahydroabietyl alcohol, Tetrahydroabietyl Alcohol
IUPAC-namn (1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,7,8,8a,9,10,10a-dodecahydrophenanthren-1-yl)methanol
LEDER CC(C)C1CCC2C(CCC3C(C)(CO)CCCC23C)C1
Specifikationer
Kemiskt namn eller material Tetrahydroabietyl Alcohol
Kvantitet 5 mg
InChI formel InChI=1S/C20H36O/c1-14(2)15-6-8-17-16(12-15)7-9-18-19(3,13-21)10-5-11-20(17,18)4/h14-18,21H,5-13H2,1-4H3
Formel vikt 292.2766