Läs mer
Tetrahydrofuran-2,5-dicarboxylic Acid (>80%), TRC
Produktkod. 30417144
Change view
Kvantitet:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417144 500 mg
30411174 1 g
30412432 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417144 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT222765500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6338-43-8
Molekylformel C6 H8 O5
Molekylvikt (g/mol) 160.12
Synonym 2,5-Anhydro-3,4-dideoxyhexaric acid (ACI), Hexaric acid, 2,5-anhydro-3,4-dideoxy- (9CI, ACI), 2,5-Anhydro-3,4-dideoxyhexaric acid (ACI), 2,5-Furandicarboxylic acid, tetrahydro- (6CI, 7CI, 8CI), NSC 40743, Tetrahydrofuran-2,5-dicarboxylic acid
IUPAC-namn oxolane-2,5-dicarboxylic acid
LEDER OC(=O)C1CCC(O1)C(=O)O
Specifikationer
Kemiskt namn eller material Tetrahydrofuran-2,5-dicarboxylic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C6H8O5/c7-5(8)3-1-2-4(11-3)6(9)10/h3-4H,1-2H2,(H,7,8)(H,9,10)
Formel vikt 160.0372