Läs mer
Tetrahydrofuran-2,5-dicarboxylic Acid, TRC
Produktkod. 30415480
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30415480 500 mg
1 options
Produktkod. 30415480 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT293525500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetrahydrofuran-2,5-dicarboxylic Acid
CAS 6338-43-8
Molekylformel C6 H8 O5
Kvantitet 500 mg
Synonym 2,5-Anhydro-3,4-dideoxyhexaric acid (ACI), Hexaric acid','2,5-anhydro-3,4-dideoxy- (9CI','ACI), 2,5-Anhydro-3,4-dideoxyhexaric acid (ACI), 2,5-Furandicarboxylic acid','tetrahydro- (6CI','7CI','8CI), NSC 40743, Tetrahydrofuran-2,5-dicarboxylic acid
InChI formel InChI=1S/C6H8O5/c7-5(8)3-1-2-4(11-3)6(9)10/h3-4H,1-2H2,(H,7,8)(H,9,10)
LEDER OC(=O)C1CCC(O1)C(=O)O
IUPAC-namn oxolane-2,5-dicarboxylic acid
Molekylvikt (g/mol) 160.12
Formel vikt 160.0372