Learn More
Tetrahydrofuran, Honeywell Burdick & Jackson™ Biosyn™
Catalog Number p-7203052
Click to view available options
Packaging:
Glass Bottle
Quantity:
2.5 L
4 x 2.5 L
72.11
72.11
For biosynthesis and applications that require low water content. Contains BHT, > 99.9% purity.
  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY
pH Neutral
Melting Point -108°C
Color Colorless
UN Number UN2056
Percent Purity >99.9%
Density 0.89 g/cm3 (20°C)
Boiling Point 65°C (1013 hPa)
Quantity 2.5 L
Explosion Limit 1.5% to 12% (V)
Flash Point -21°C
Packaging Glass Bottle
Physical Form Liquid
Product Line BioSyn™
Vapor Pressure 200 hPa (20°C)
Viscosity 0.48 mPa.s (20°C)