missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Biograde, 99.8%, unstab., Thermo Scientific Chemicals

Catalog Number p-7059392
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

Nucleic acid and peptide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -108°C
Color Colorless
Density 0.889 g/mL
Boiling Point 66°C
Quantity 1 L
Flash Point −17°C (1°F)
Odor Ether-like
Physical Form Liquid
Refractive Index 1.407
UN Number UN2056
Beilstein 102391
Merck Index 14,9211
Solubility Information Miscible with water.
Formula Weight 72.11
Percent Purity 99.8%
Grade Biograde
Chemical Name or Material Tetrahydrofuran, Unstabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'