missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran in Dimethyl Sulfoxide, SPEX CertiPrep™

Catalog Number 16379757
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Tetrahydrofuran109-99-93600μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Physical Form Fluid
CAS 109-99-9
Molecular Formula C4H8O
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
PubChem CID 8028
ChEBI CHEBI:26911
Explosion Limit 1.8% (volume); 63% (volume)
Grade USP
Concentration or Composition (by Analyte or Components) 3600μg/mL
Ignition Point 270°C
Packaging 2mL amber prescored glass ampule
Vapor Pressure 2.5 hPA (20°C)
Analyte or Component Names Tetrahyrdofuran
Density 1.1g/cm3
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Tetrahydrofuran in Dimethyl Sulfoxide, USP 467 Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'