missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Ultra Low water, No Preservative, BakerDRY™, J.T. Baker™

Catalog Number p-5391034
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

Specifically manufactured for use in organometallic and oligonucleotide synthesis

    For Use in Organic Synthesis, including Organometallic Synthesis
Formula Weight 72.11
Grade BakerDRY™
Quantity 1 L
Flash Point −14.00°C
Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Specific Gravity 1L = 0.88kg
Chemical Name or Material Tetrahydrofuran, Ultra Low water
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'