missing translation for 'onlineSavingsMsg'
Läs mer

Tetrahydrofurfuryl Alcohol, Sigma-Aldrich™

Produktkod. 15831781
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 L
Förpackningsstorlek:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15831781 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Produktkod. 15831781 Leverantör Sigma Aldrich Leverantörsnummer 8223072500
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Tetrahydrofurfuryl Alcohol
Smältpunkt <-80°C
CAS 97-99-4
Densitet 1.05 g/cm3 at 20°C
Kvantitet 2.5 L
MDL-nummer MFCD00005372
Synonym Tetrahydrofurfuryl alcohol, Tetrahydrofurancarbinol, 2-Hydroxymethyltetrahydrofuran, Tetrahydrofurfuryl alcohol
Kokpunkt 178°C/1013 hPa
Molekylvikt (g/mol) 102.13
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.