Läs mer
Tetrahydroharmine, Mikromol™
Produktkod. 17907964
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17907964 25 mg
17927764 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17907964 Leverantör Mikromol Leverantörsnummer MM2562.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tetrahydroharmine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tetrahydro-Harmine
CAS 17019-01-1
Analyt- eller komponentnamn tetrahydro-Harmine
Frakt skick Room Temperature
Molekylformel C13 H16 N2 O
Certifikat och efterlevnad ISO 9001
Synonym 1H-Pyrido[3,4-b]indole, 2,3,4,9-tetrahydro-7-methoxy-1-methyl-, Harmine, tetrahydro- (6CI), 1,2,3,4-Tetrahydroharmine, Leptaflorine, Tetrahydroharmine, 1,2-Dihydroharmaline, (+/-)-Tetrahydroharmine
InChI formel InChI=1S/C13H16N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-4,7-8,14-15H,5-6H2,1-2H3
LEDER COc1ccc2c3CCNC(C)c3[nH]c2c1
IUPAC-namn 7-methoxy-1-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
Molekylvikt (g/mol) 216.28
Kvantitet 25 mg
Formel vikt 216.1263 g/mol