Läs mer
Tetrahydroharmine, TRC
Produktkod. 30450319
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30450319 25 mg
1 options
Produktkod. 30450319 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT31028525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material tetrahydro-Harmine
CAS 17019-01-1
Anteckningar om renhetsgrad HPLC
Molekylformel C13 H16 N2 O
Kvantitet 25 mg
Synonym 1H-Pyrido[3,4-b]indole','2,3,4,9-tetrahydro-7-methoxy-1-methyl-, Harmine','tetrahydro- (6CI), (±)-Tetrahydroharmine, 1,2,3,4-Tetrahydroharmine, Leptaflorine, Tetrahydroharmine, 1,2-Dihydroharmaline
InChI formel InChI=1S/C13H16N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-4,7-8,14-15H,5-6H2,1-2H3
LEDER COc1ccc2c3CCNC(C)c3[nH]c2c1
IUPAC-namn 7-methoxy-1-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
Molekylvikt (g/mol) 216.28
Formel vikt 216.1263