Läs mer
Tetrahydrophthalic Acid, TRC
Produktkod. 30452612
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452612 500 mg
30450005 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452612 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT889826500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,2,3,6-Tetrahydrophthalic acid
CAS 88-98-2
Molekylformel C8 H10 O4
Kvantitet 500 mg
Synonym 1,2,3,6-Tetrahydrophthalic acid, HN 200, NSC 239116, Rikacid TH-W, Tetrahydrophthalic acid, Δ4-Tetrahydrophthalic acid, THPA
InChI formel InChI=1S/C8H10O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-2,5-6H,3-4H2,(H,9,10)(H,11,12)
LEDER OC(=O)C1CC=CCC1C(=O)O
IUPAC-namn cyclohex-4-ene-1,2-dicarboxylic acid
Molekylvikt (g/mol) 170.16
Formel vikt 170.0579