Learn More
Tetrahydrothiopyran, 98%, Thermo Scientific Chemicals
Catalog Number p-7043848
Click to view available options
Quantity:
1 g
5 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1613-51-0
C5H10S
102.195
MFCD00006662
YPWFISCTZQNZAU-UHFFFAOYSA-N
pentamethylene sulfide, tetrahydrothiopyran, tetrahydro-2h-thiopyran, thiacyclohexane, 2h-thiopyran, tetrahydro, penthiophane, tetrahydrothiapyran, tetrahydrothiopyrane, thian, thian-4-yl
15367
thiane
C1CCSCC1
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1613-51-0
C5H10S
102.195
MFCD00006662
YPWFISCTZQNZAU-UHFFFAOYSA-N
pentamethylene sulfide, tetrahydrothiopyran, tetrahydro-2h-thiopyran, thiacyclohexane, 2h-thiopyran, tetrahydro, penthiophane, tetrahydrothiapyran, tetrahydrothiopyrane, thian, thian-4-yl
15367
thiane
C1CCSCC1
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful 5 c synthon for the synthesis of 3-cyclopentenones. It is a organic sulfide, that has shown to have antioxidant activity.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY
Density 0.988
Boiling Point 140°C to 142°C
Flash Point 21°C (69°F)
Odor Unpleasant
Refractive Index 1.506
Quantity 1 g
UN Number UN1993
Beilstein 102437
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 102.2
Percent Purity 98%
Chemical Name or Material Tetrahydrothiopyran