missing translation for 'onlineSavingsMsg'
Learn More

Tetraisobutyldialuminoxane, 10 wt.% solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4229332
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
998-00-5, 108-88-3
C16H36Al2O
298.42
CTNFGBKAHAEKFE-UHFFFAOYSA-N
aluminum, tetrakis 2-methylpropyl-.mu.-oxodi, tetrakis isobutyl-mu-oxodialuminium, tetraisobutyl dialumoxane, aluminum, tetrakis 2-methylpropyl-mu-oxodi, bis diisobutylaluminum oxide, bis-diisobutylaluminum-oxide, tetraisobutyldialuminoxane solution, tetraisobutyldialuminoxane solution, 10 wt. % in toluene, bis 2-methylpropyl alumanyl oxy bis 2-methylpropyl alumane
16684086
bis(2-methylpropyl)alumanyloxy-bis(2-methylpropyl)alumane
CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C
This item is not returnable. View return policy
998-00-5, 108-88-3
C16H36Al2O
298.42
CTNFGBKAHAEKFE-UHFFFAOYSA-N
aluminum, tetrakis 2-methylpropyl-.mu.-oxodi, tetrakis isobutyl-mu-oxodialuminium, tetraisobutyl dialumoxane, aluminum, tetrakis 2-methylpropyl-mu-oxodi, bis diisobutylaluminum oxide, bis-diisobutylaluminum-oxide, tetraisobutyldialuminoxane solution, tetraisobutyldialuminoxane solution, 10 wt. % in toluene, bis 2-methylpropyl alumanyl oxy bis 2-methylpropyl alumane
16684086
bis(2-methylpropyl)alumanyloxy-bis(2-methylpropyl)alumane
CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8670g/mL
Boiling Point 111°C
Flash Point -8°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.867
Formula Weight 298.42
Physical Form Solution
Chemical Name or Material Tetraisobutyldialuminoxane
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetraisobutyldialuminoxane, 10 wt.% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'