Läs mer
Tetrakis(acetonitrile)copper(I) Hexafluorophosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tetrakis(acetonitrile)copper(I) Hexafluorophosphate |
| CAS | 64443-05-6 |
| Molekylformel | C8H12CuF6N4P |
| Kvantitet | 500 mg |
| Synonym | Copper(1+)','tetrakis(acetonitrile)-','(T-4)-','hexafluorophosphate(1-) (9CI), Phosphate(1-)','hexafluoro-','(T-4)-tetrakis(acetonitrile)copper(1+) (ZCI), Copper(I) tetrakis(acetonitrile) hexafluorophosphate, Cuprous tetrakis(acetonitrile) hexafluorophosphate, Tetra(acetonitrile)copper(1+) hexafluorophosphate, Tetrakis(acetonitrile)copper hexafluorophosphate, Tetrakis(acetonitrile)copper(1+) hexafluorophosphate, Tetrakis(acetonitrile)copper(1+) hexafluorophosphate(1-), Tetrakis(acetonitrile)copper(I) hexafluorophosphate |
| InChI formel | InChI=1S/4C2H3N.Cu.F6P/c4*1-2-3;;1-7(2,3,4,5)6/h4*1H3;;/q;;;;+1;-1 |
| LEDER | [F-][P+5]([F-])([F-])([F-])([F-])[F-].CC#[N][Cu+]([N]#CC)([N]#CC)[N]#CC |
| Molekylvikt (g/mol) | 372.72 |
| Formel vikt | 372 |