Läs mer
Tetramethylammonium Bicarbonate (60% in water), TRC
Produktkod. 30451255
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30451255 500 mg
30449191 5 g
30450323 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30451255 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT890223500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58345-96-3
Molekylformel C5H13NO3
Molekylvikt (g/mol) 135.16
Synonym Tetramethylammonium Hydrogen Carbonate
IUPAC-namn hydrogen carbonate;tetramethylazanium
LEDER C[N+](C)(C)C.OC(=O)[O-]
Specifikationer
Kemiskt namn eller material Tetramethylammonium Bicarbonate
Kvantitet 500 mg
InChI formel InChI=1S/C4H12N.CH2O3/c1-5(2,3)4;2-1(3)4/h1-4H3;(H2,2,3,4)/q+1;/p-1
Formel vikt 135.0895
Koncentration eller sammansättning (efter analyt eller komponenter) 60%