Läs mer
Tetramethylol Acetylenediurea, TRC
Produktkod. 30132407
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132407 5 g
30132848 10 g
30008477 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132407 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8897085G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5395-50-6
Molekylformel C8 H14 N4 O6
Molekylvikt (g/mol) 262.22
Synonym Glycoluril, 1,3,4,6-tetrakis(hydroxymethyl)- (6CI,7CI,8CI), Tetrahydro-1,3,4,6-tetrakis(hydroxymethyl)imidazo[4,5-d]imidazole-2,5(1H,3H)-dione, 1,3,4,6-Tetrakis(hydroxymethyl)glycoluril, 2,4,6,8-Tetrakis(hydroxymethyl)glycoluril, 2,5-Dioxo-1,3,4,6-tetrakis(hydroxymethyl)hexahydroimidazo[4,5-d]imidazole, Cymel 1172, Fixapret 140, N,N',N' ',N' ' '-Tetrakis(hydroxymethyl)glycoluril, NSC 3319, NSC 67452, Protectol TD, Tetrahydro-1,3,4,6-tetrakis(hydroxymethyl)imidazo[4,5-d]imidazol-2,5(1H,3H)-dione, Tetrakis(hydroxymethyl)acetylenediurea, Tetrakis(hydroxymethyl)acetyleneurea, Tetrakis(hydroxymethyl)glycoluril, Tetramethylol acetylene carbamide, Tetramethylolacetylenediurea, Tetramethylolglycoluril
IUPAC-namn 1,3,4,6-tetrakis(hydroxymethyl)-3a,6a-dihydroimidazo[4,5-d]imidazole-2,5-dione
LEDER OCN1C2C(N(CO)C(=O)N2CO)N(CO)C1=O
Specifikationer
Kemiskt namn eller material Tetrakis(hydroxymethyl)glycoluril
Kvantitet 5 g
InChI formel InChI=1S/C8H14N4O6/c13-1-9-5-6(11(3-15)7(9)17)12(4-16)8(18)10(5)2-14/h5-6,13-16H,1-4H2
Formel vikt 262.0913