Läs mer
Tetramethylphosphorodiamidic Fluoride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimefox |
| CAS | 115-26-4 |
| Molekylformel | C4 H12 F N2 O P |
| Kvantitet | 50 mg |
| Synonym | Phosphorodiamidic fluoride','tetramethyl- (6CI,7CI,8CI,9CI), N,N,N',N'-Tetramethylphosphorodiamidic fluoride, BFPO, Bis(dimethylamido)fluorophosphate, Bis(dimethylamido)fluorophosphine oxide, Bis(dimethylamido)phosphoryl fluoride, Bis(dimethylamino)fluorophosphine oxide, Bis(dimethylamino)phosphorofluoridate, CR 409, DIFO, Dimefox, Fluorophosphoric acid di(dimethylamide), Hanane, Pestox 14, Phosphine oxide','bis(dimethylamino)fluoro-, Terra-Sytam, Tetramethyldiamidophosphoric fluoride, Tetramethylphosphorodiamidic fluoride |
| InChI formel | InChI=1S/C4H12FN2OP/c1-6(2)9(5,8)7(3)4/h1-4H3 |
| LEDER | CN(C)P(=O)(F)N(C)C |
| IUPAC-namn | N-[dimethylamino(fluoro)phosphoryl]-N-methylmethanamine |
| Molekylvikt (g/mol) | 154.12 |
| Formel vikt | 154.0671 |