Learn More
Tetramethylsilane, for the calibration of NMR spectra for NMR spectroscopy, MagniSolv™, MilliporeSigma™
Product Code. 15893871
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
Product Code. Quantity Packaging Unit Size
15893871 100 mL Glass Bottle 100mL
1 options
Product Code. 15893871 Supplier Sigma Aldrich Supplier No. 1081830100
 Restricted Item
Beräknad leverans: 18-11-2026
For NMR Spectroscopy
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Odor Characteristic
Packaging Glass Bottle
Vapor Pressure 750 hPa at 20°C (68°F)
Quantity 100 mL
CAS 75-76-3
Molecular Formula C4H12Si
MDL Number MFCD00008274
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
SMILES C[Si](C)(C)C
IUPAC Name tetramethylsilane
Molecular Weight (g/mol) 88.23
PubChem CID 6396
ChEBI CHEBI:85361
Formula Weight 88.23
Chemical Name or Material Tetramethylsilane