Läs mer
Tetramethyluric Acid, TRC
Produktkod. 30450256
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30450256 1 g
1 options
Produktkod. 30450256 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3056601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tetramethyluric Acid
CAS 2309-49-1
Molekylformel C9H12N4O3
Kvantitet 1 g
Synonym 1,3,7,9-Tetramethyluric Acid, Ba 2750, NSC 8809, Temorine, Temurin, Tetramethyluric Acid, Theacrine, 7,9-Dihydro-1,3,7,9-tetramethyl-1H-purine-2,6,8(3H)-trione
InChI formel InChI=1S/C9H12N4O3/c1-10-5-6(11(2)8(10)15)12(3)9(16)13(4)7(5)14/h1-4H3
LEDER CN1C(=O)N(C)C2=C(N(C)C(=O)N2C)C1=O
IUPAC-namn 1,3,7,9-tetramethylpurine-2,6,8-trione
Molekylvikt (g/mol) 224.22
Formel vikt 224.0909