missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylcyclopentadienone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It is an easily made building block for many organic and organometallic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an easily made building block for many organic and organometallic compounds.
Solubility
Soluble in Benzene. Insoluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
It is an easily made building block for many organic and organometallic compounds.
Solubility
Soluble in Benzene. Insoluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 217°C to 219°C |
Quantity | 5 g |
Beilstein | 683115 |
Solubility Information | Soluble in Benzene. Insoluble in water. |
Formula Weight | 384.48 |
Percent Purity | 98% |
Chemical Name or Material | Tetraphenylcyclopentadienone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'