missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-199163
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2001-45-8
C24H20ClP
374.85
MFCD00011916
WAGFXJQAIZNSEQ-UHFFFAOYSA-M
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
164911
tetraphenylphosphanium;chloride
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
2001-45-8
C24H20ClP
374.85
MFCD00011916
WAGFXJQAIZNSEQ-UHFFFAOYSA-M
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
164911
tetraphenylphosphanium;chloride
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Melting Point 275.0°C to 280.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C6H5)4PCl
Quantity 10 g
Beilstein 16, II, 373
Solubility Information Solubility in water: soluble.
Formula Weight 374.84
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylphosphonium chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tetraphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'