Läs mer
Tetrasul, TRC
Produktkod. 30450603
Change view
Kvantitet:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450603 1 g
30450994 25 mg
30451812 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450603 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3064401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2227-13-6
Molekylformel C12 H6 Cl4 S
Molekylvikt (g/mol) 324.05
Synonym Sulfide, p-chlorophenyl 2,4,5-trichlorophenyl (6CI,7CI,8CI), 1,2,4-Trichloro-5-[(4-chlorophenyl)thio]benzene, 2,4,4',5-Tetrachlorodiphenyl sulfide, 4-Chlorophenyl 2,4,5-trichlorophenyl sulfide, Animert, Animert V 101, Tetrasul, V 101, p-Chlorophenyl 2,4,5-trichlorophenyl sulfide
IUPAC-namn 1,2,4-trichloro-5-(4-chlorophenyl)sulfanylbenzene
LEDER Clc1ccc(Sc2cc(Cl)c(Cl)cc2Cl)cc1
Specifikationer
Kemiskt namn eller material Tetrasul
Kvantitet 1 g
InChI formel InChI=1S/C12H6Cl4S/c13-7-1-3-8(4-2-7)17-12-6-10(15)9(14)5-11(12)16/h1-6H
Formel vikt 321.8944