missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAM Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22586-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.Specifications
TFAM | |
Polyclonal | |
Unconjugated | |
TFAM | |
MtTF1, mtTFA, TCF 6, TCF6, TCF6L1, TCF6L2, TCF6L3, TFAM, Transcription factor 6, Transcription factor 6 like 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21780, 7019 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40630, Q00059 | |
TFAM | |
TFAM Fusion Protein Ag18299 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |