Learn More
TFAP, MedChemExpress
Product Code. 16467407
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467407 5 mg 5mg
16445526 100 mg 100mg
16424698 10 mg 10mg
16410987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16467407 Supplier Medchem Express Supplier No. HY112731
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress TFAP is a selective cyclooxygenase-1 (COX-1) inhibitor, with an IC50 of 0.8 μM.
TFAP is a selective cyclooxygenase-1 (COX-1) inhibitor, with an IC50 of 0.8 μM.
Specifications
Chemical Name or Material TFAP
CAS 1011244-68-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H10F3N3O
Quantity 5 mg
Synonym N-(5-Aminopyridin-2-yl)-4-(trifluoromethyl)benzamide
Solubility Information DMSO : ≥ 155 mg/mL (551.15 mM)
SMILES O=C(NC1=NC=C(N)C=C1)C2=CC=C(C(F)(F)F)C=C2
Molecular Weight (g/mol) 281.23
Formula Weight 281.23
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism