missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12017-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNAbinding, and HLH and ZIP domains, which are used foroligomerization. Transcription factor AP4 activates both viral andcellular genes by binding to the symmetrical DNA sequence CAGCTG.Specifications
TFAP4 | |
Polyclonal | |
Unconjugated | |
TFAP4 | |
AP 4, bHLHc41, TFAP4, Transcription factor AP 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360482, 7023, 83383 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q01664 | |
Tfap4 | |
TFAP4 Fusion Protein Ag2633 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |