missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TFAP4 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879473
|
Proteintech
12017-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16869473
|
Proteintech
12017-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNAbinding, and HLH and ZIP domains, which are used foroligomerization. Transcription factor AP4 activates both viral andcellular genes by binding to the symmetrical DNA sequence CAGCTG.Specifications
TFAP4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q01664 | |
360482, 7023, 83383 | |
TFAP4 Fusion Protein Ag2633 | |
Primary | |
-20°C | |
TFAP4 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AP 4, bHLHc41, TFAP4, Transcription factor AP 4 | |
Tfap4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |