Learn More
WTW™ TFK™ 325/HC-3 Temperature Sensor
Product Code. 10727602
Change view
Click to view available options
Length (Metric) Cable:
3 m
Unit Size:
Each
Product Code. Length (Metric) Cable Unit Size
10727602 3 m Each
1 options
Product Code. 10727602 Supplier WTW™ Supplier No. 109823
Beräknad leverans: 22-12-2026
Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor
pH and ORP measurements must always have a temperature indication. During pH measurement, the indication of a temperature to compensate the temperature effect, and the documentation, with ORP measurement, only for documentation purposes. During ISE measurement it is important that the calibration and the measurement are performed in the same temperature range. Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor. The temperature sensors are equipped with two 4 mm banana plugs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For pH electrodes
Type Temperature Sensor
Length (Metric) 3 m
Length (Metric) Cable 3 m
Temperature Sensor Yes