missing translation for 'onlineSavingsMsg'
Läs mer

TfR (Transferrin R) Antibody (8D3) - Azide and BSA Free, Novus Biologicals™

Produktkod. 18028895 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18028895 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18028895 Leverantör Novus Biologicals Leverantörsnummer NB100649790.05MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rat Monoclonal Antibody

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, In vivo assay, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen TfR (Transferrin R)
Användningsområden Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Klassificering Monoclonal
Klona 8D3
Koncentration 1.0 mg/mL
Konjugera Unconjugated
Utspädning Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, In vivo assay
Formulering PBS with No Preservative
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Gensymboler TFRC
Värd art Rat
Immunogen Mouse transformed endothelioma cell line t.end1.
Reningsmetod Protein G purified
Kvantitet 0.05 mg
Regulatorisk status RUO
Forskningsdisciplin Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primär eller sekundär Primary
Gen-ID (Entrez) 7037
Testspecificitet NB100-64979 recognizes native, soluble and denatured forms of murine CD71. The antibody has been used as a BBB transporter vector in mice and is suitable for studying CD71 expression, and iron uptake into different tissues, in the mouse.
Målarter Mouse
Innehåll och lagring Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotyp IgG2a
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.