Läs mer
Tocris Bioscience™ TG 003
GREENER_CHOICE
Produktkod. 18094952
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18094952 50 mg
18054852 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18094952 Leverantör Tocris Bioscience™ Leverantörsnummer 4336/50
Potent inhibitor of Clk-family kinases; also inhibits DYRK1A/B
Potent, ATP-competitive inhibitor of Clk-family kinases (IC50 values are 15, 20 and 200 nM for mClk4, 1 and 2 respectively and >10 μM for mClk3). Also inhibits DYRK1A/B (IC50 values are 24 and 34 nM respectively). Suppresses serine/arginine-rich protein phosphorylation; affects the regulation of alternative splicing by phosphorylation of SR protein both in vitro and in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 719277-26-6
Molekylformel C13H15NO2S
Specifikationer
Kemiskt namn eller material TG 003
Kvantitet 50 mg
Formel vikt 249.33
Renhet >98%
Molekylvikt (g/mol) 249.328