Learn More
Invitrogen™ TGF beta-2 Human ELISA Kit
Sandwich ELISA Kit
6020.00 SEK - 27960.00 SEK
Specifications
Accession Number | P61812 |
---|---|
Assay Sensitivity | 15 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425143
|
Invitrogen™
EHTGFB2 |
96 Tests |
6020.00 SEK
1 set |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17175569
|
Invitrogen™
EHTGFB2X5 |
5 x 96 Tests |
27960.00 SEK
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
The Human Transforming Growth Factor beta2 (TGFβ2) ELISA quantitates Hu TGFβ2 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TGFβ2. Principle of the method The Human TGFβ2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Human Transforming Growth Factor beta 2 (TGF beta 2) is a member of a superfamily of disulfide-linked homodimeric proteins secreted as latent proteins and stored at the cell surface and extracellular matrix. The bioactive TGF beta 2 is released from a latent complex by proteolytic processing and conformation changes. TGF beta 2 regulates cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix and is also involved in embryogenesis, tissue remodeling and wound healing.Specifications
P61812 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
G-TSF,LDS4,TGF-beta2 | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
15 pg/mL | |
Biotin | |
Cytokines & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP- 2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta1, TGF-beta3, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
7042 | |
<12% | |
HRP | |
Angiogenesis | |
LDS4, TGF-beta2 | |
1 hr. 20 min. |